Product Details

SNP ID
rs118145687
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12834601 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACATCCGCCCCCCCACCGCGATC[A/C]TCAAGTCCAGCCCAGCTTGTGGGTG
Phenotype
MIM: 612256 MIM: 609553
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MAST1 PubMed Links

Gene Details

Gene
MAST1
Gene Name
microtubule associated serine/threonine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014975.2 159 Intron NP_055790.1
XM_011527805.2 159 Intron XP_011526107.1
XM_011527808.2 159 Intron XP_011526110.1
XM_017026501.1 159 Intron XP_016881990.1
Gene
RTBDN
Gene Name
retbindin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080997.2 159 Intron NP_001074466.1
NM_001270440.1 159 Intron NP_001257369.1
NM_001270441.1 159 Intron NP_001257370.1
NM_001270442.1 159 Intron NP_001257371.1
NM_001270443.1 159 Intron NP_001257372.1
NM_001270444.1 159 Intron NP_001257373.1
NM_001270445.1 159 Intron NP_001257374.1
NM_031429.2 159 Intron NP_113617.1
XM_005260089.1 159 Intron XP_005260146.1
XM_005260090.3 159 UTR 5 XP_005260147.1
XM_011528343.1 159 Intron XP_011526645.1
XM_011528344.2 159 Intron XP_011526646.1
XM_011528345.1 159 Intron XP_011526647.1
XM_017027343.1 159 Intron XP_016882832.1
XM_017027344.1 159 Intron XP_016882833.1

View Full Product Details