Product Details

SNP ID
rs112017208
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14841081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCAGCACCTGTGTGTGAATTG[C/T]GGGTGGCAGCAGAACTAAGGTACAC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR7A10 PubMed Links
Additional Information
For this assay, SNP(s) [rs3752194,rs3752195] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
OR7A10
Gene Name
olfactory receptor family 7 subfamily A member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005190.1 797 Missense Mutation CAC,CGC H266R NP_001005190.1
Gene
OR7A5
Gene Name
olfactory receptor family 7 subfamily A member 5
There are no transcripts associated with this gene.

Gene
OR7C1
Gene Name
olfactory receptor family 7 subfamily C member 1
There are no transcripts associated with this gene.

View Full Product Details