Product Details

SNP ID
rs112660293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18193350 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGCAGCTTCTATTCCAGGGCCG[C/G]GCGCTGCTCGTCACTAACACGCTGG
Phenotype
MIM: 616133 MIM: 179490
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MPV17L2 PubMed Links

Gene Details

Gene
MPV17L2
Gene Name
MPV17 mitochondrial inner membrane protein like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032683.2 121 Silent Mutation CGC,CGG R23R NP_116072.2
Gene
RAB3A
Gene Name
RAB3A, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details