Product Details
- SNP ID
-
rs114090481
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:63559929 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAAGAGCCCAGCCCCGCCCTCACC[A/G]ATCAGGCAGAGCCCCTCCTGGGCCC
- Phenotype
-
MIM: 611265
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FNDC11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3810477] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FNDC11
- Gene Name
- fibronectin type III domain containing 11
There are no transcripts associated with this gene.
- Gene
- HELZ2
- Gene Name
- helicase with zinc finger 2
View Full Product Details