Product Details

SNP ID
rs112926360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:56001785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCCTAGTTCTATTGGTTATAAA[A/G]TTAACTCTAAATTTTAATAAGTGAC
Phenotype
MIM: 615029
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CBLN4 PubMed Links

Gene Details

Gene
CBLN4
Gene Name
cerebellin 4 precursor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080617.5 Intron NP_542184.1

View Full Product Details