Product Details

SNP ID
rs112774015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36545073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGACATGCTGGCCTCGCTGGGTGA[A/G]CTGGGACAGGGACAGGGGTGAGATG
Phenotype
MIM: 609905
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DLGAP4-AS1 PubMed Links

Gene Details

Gene
DLGAP4-AS1
Gene Name
DLGAP4 antisense RNA 1
There are no transcripts associated with this gene.

Gene
MYL9
Gene Name
myosin light chain 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006097.4 Intron NP_006088.2
NM_181526.2 Intron NP_852667.1

View Full Product Details