Product Details

SNP ID
rs118044390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:38728135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGGGCCTCTTCGCGCTCGTCGC[C/T]TACCTCACCTGGGCCGACGAGACCA
Phenotype
MIM: 616440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC32A1 PubMed Links

Gene Details

Gene
SLC32A1
Gene Name
solute carrier family 32 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080552.2 1337 Silent Mutation GCC,GCT A358A NP_542119.1

View Full Product Details