Product Details

SNP ID
rs115726903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2482229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCATTCTCCCCACACATGCAGAA[A/T]TTCTATTCAATATACTCTGGGGCTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF343 PubMed Links

Gene Details

Gene
ZNF343
Gene Name
zinc finger protein 343
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282495.1 3084 UTR 3 NP_001269424.1
NM_001282496.1 3084 UTR 3 NP_001269425.1
NM_001282497.1 3084 UTR 3 NP_001269426.1
NM_001282498.1 3084 UTR 3 NP_001269427.1
NM_001282499.1 3084 Intron NP_001269428.1
NM_001321800.1 3084 UTR 3 NP_001308729.1
NM_001321801.1 3084 UTR 3 NP_001308730.1
NM_001321802.1 3084 UTR 3 NP_001308731.1
NM_001321803.1 3084 UTR 3 NP_001308732.1
NM_001321805.1 3084 UTR 3 NP_001308734.1
NM_024325.5 3084 UTR 3 NP_077301.4
XM_017028062.1 3084 UTR 3 XP_016883551.1

View Full Product Details