Product Details

SNP ID
rs113647026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41309748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGACCAGTTTAAAAGCTTAGAC[A/G]CATTGATCTGAATATTTGATGGCTG
Phenotype
MIM: 608617
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM3B PubMed Links

Gene Details

Gene
FAM3B
Gene Name
family with sequence similarity 3 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058186.3 Intron NP_478066.3
NM_206964.1 Intron NP_996847.1
XM_011529648.2 Intron XP_011527950.2
XM_011529649.2 Intron XP_011527951.1

View Full Product Details