Product Details

SNP ID
rs111668637
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44612170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCATGTCTGTCTGCTCCAGTGAC[A/G]TGGGCCATGTCAGCCGAGTCTCCTC
Phenotype
MIM: 612920
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KRTAP10-7 PubMed Links
Additional Information
For this assay, SNP(s) [rs411254] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KRTAP10-7
Gene Name
keratin associated protein 10-7
There are no transcripts associated with this gene.

Gene
KRTAP10-8
Gene Name
keratin associated protein 10-8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198695.2 92 Missense Mutation ATG,GTG M24V NP_941968.2
Gene
TSPEAR
Gene Name
thrombospondin type laminin G domain and EAR repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272037.1 92 Intron NP_001258966.1
NM_144991.2 92 Intron NP_659428.2

View Full Product Details