Product Details

SNP ID
rs117966061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:46329827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAAAAACCAAAAGCTGGTAAAA[G/T]GTAAGGATACTTTCTCTAGGAAAAG
Phenotype
MIM: 605925
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C21orf58 PubMed Links

Gene Details

Gene
C21orf58
Gene Name
chromosome 21 open reading frame 58
There are no transcripts associated with this gene.

Gene
PCNT
Gene Name
pericentrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001315529.1 Intron NP_001302458.1
NM_006031.5 Intron NP_006022.3
XM_005261124.4 Intron XP_005261181.1
XM_011529594.2 Intron XP_011527896.1
XM_017028362.1 Intron XP_016883851.1
XM_017028363.1 Intron XP_016883852.1

View Full Product Details