Product Details

SNP ID
rs114774816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19523062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATTTACTAAGCAGATTCTTAGC[C/T]TTCCCACTCCCGCCCTCTCTCAAGC
Phenotype
MIM: 603465 MIM: 602101
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC45 PubMed Links

Gene Details

Gene
CDC45
Gene Name
cell division cycle 45
There are no transcripts associated with this gene.

Gene
CLDN5
Gene Name
claudin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130861.1 1664 UTR 3 NP_001124333.1
NM_003277.3 1664 UTR 3 NP_003268.2
XM_017028929.1 1664 UTR 3 XP_016884418.1
XM_017028930.1 1664 UTR 3 XP_016884419.1

View Full Product Details