Product Details

SNP ID
rs112469158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19523845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGGACGACAATGTTGGCGAACCA[A/G]CAGAGTGGCACGAGCGCCAGCAGCC
Phenotype
MIM: 603465 MIM: 602101
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC45 PubMed Links

Gene Details

Gene
CDC45
Gene Name
cell division cycle 45
There are no transcripts associated with this gene.

Gene
CLDN5
Gene Name
claudin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130861.1 881 Silent Mutation TGC,TGT C222C NP_001124333.1
NM_003277.3 881 Silent Mutation TGC,TGT C222C NP_003268.2
XM_017028929.1 881 Silent Mutation TGC,TGT C222C XP_016884418.1
XM_017028930.1 881 Silent Mutation TGC,TGT C222C XP_016884419.1

View Full Product Details