Product Details
- SNP ID
-
rs115897309
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:30555514 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCGACGCTCCACCTCCAGGATGTG[C/T]TCCTGCACCTGCGGGCTGCTGGGGT
- Phenotype
-
MIM: 602300
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GAL3ST1
PubMed Links
Gene Details
- Gene
- GAL3ST1
- Gene Name
- galactose-3-O-sulfotransferase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001318103.1 |
885 |
Silent Mutation |
GAA,GAG |
E237E |
NP_001305032.1 |
NM_001318104.1 |
885 |
Silent Mutation |
GAA,GAG |
E237E |
NP_001305033.1 |
NM_001318105.1 |
885 |
Silent Mutation |
GAA,GAG |
E237E |
NP_001305034.1 |
NM_001318106.1 |
885 |
Silent Mutation |
GAA,GAG |
E237E |
NP_001305035.1 |
NM_001318107.1 |
885 |
Silent Mutation |
GAA,GAG |
E238E |
NP_001305036.1 |
NM_001318108.1 |
885 |
Silent Mutation |
GAA,GAG |
E237E |
NP_001305037.1 |
NM_001318109.1 |
885 |
Silent Mutation |
GAA,GAG |
E262E |
NP_001305038.1 |
NM_001318110.1 |
885 |
Silent Mutation |
GAA,GAG |
E237E |
NP_001305039.1 |
NM_001318111.1 |
885 |
Silent Mutation |
GAA,GAG |
E237E |
NP_001305040.1 |
NM_001318112.1 |
885 |
Silent Mutation |
GAA,GAG |
E237E |
NP_001305041.1 |
NM_001318113.1 |
885 |
Silent Mutation |
GAA,GAG |
E237E |
NP_001305042.1 |
NM_001318114.1 |
885 |
Silent Mutation |
GAA,GAG |
E238E |
NP_001305043.1 |
NM_001318115.1 |
885 |
Silent Mutation |
GAA,GAG |
E237E |
NP_001305044.1 |
NM_001318116.1 |
885 |
Silent Mutation |
GAA,GAG |
E237E |
NP_001305045.1 |
NM_004861.2 |
885 |
Silent Mutation |
GAA,GAG |
E237E |
NP_004852.1 |
XM_011530518.2 |
885 |
Silent Mutation |
GAA,GAG |
E264E |
XP_011528820.1 |
XM_011530522.2 |
885 |
Silent Mutation |
GAA,GAG |
E238E |
XP_011528824.1 |
XM_011530524.1 |
885 |
Silent Mutation |
GAA,GAG |
E238E |
XP_011528826.1 |
XM_011530528.1 |
885 |
Silent Mutation |
GAA,GAG |
E238E |
XP_011528830.1 |
XM_017029096.1 |
885 |
Silent Mutation |
GAA,GAG |
E238E |
XP_016884585.1 |
XM_017029097.1 |
885 |
Silent Mutation |
GAA,GAG |
E238E |
XP_016884586.1 |
XM_017029098.1 |
885 |
Silent Mutation |
GAA,GAG |
E237E |
XP_016884587.1 |
- Gene
- SEC14L6
- Gene Name
- SEC14 like lipid binding 6
There are no transcripts associated with this gene.
View Full Product Details