Product Details

SNP ID
rs113417633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:83873034 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACATCTTTATACTTCCAAAACCC[A/G]TCTTAAAAAAGCAGAATATAAAAAG
Phenotype
MIM: 300768
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYLC1 PubMed Links

Gene Details

Gene
CYLC1
Gene Name
cylicin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271680.1 380 Intron NP_001258609.1
NM_021118.2 380 Missense Mutation CAT,CGT H109R NP_066941.1
XM_005262086.3 380 Missense Mutation CAT,CGT H108R XP_005262143.1

View Full Product Details