Product Details

SNP ID
rs113043096
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:143508927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTCCTCCTTTGAAGGTCCTTCA[C/G]ATGGGCCTAGGTCTTCATCCTCATT
Phenotype
MIM: 300666
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPANXN3 PubMed Links

Gene Details

Gene
SPANXN3
Gene Name
SPANX family member N3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009609.3 Intron NP_001009609.2
XM_017029264.1 Intron XP_016884753.1

View Full Product Details