Product Details

SNP ID
rs111725135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:19606073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAGGATGCAGTACTGATAAAAA[C/T]ACTGCTTTGTTTTTATCTCACCTCT
Phenotype
MIM: 400031
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TXLNGY PubMed Links

Gene Details

Gene
TXLNGY
Gene Name
taxilin gamma pseudogene, Y-linked
There are no transcripts associated with this gene.

View Full Product Details