Product Details
- SNP ID
-
rs112710920
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.Y:2979398 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTAAAGAGATGCCATTCAAGTGTG[A/G]CATTTGTCTTCTGACTTTCTCAGAT
- Phenotype
-
MIM: 490000
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZFY
PubMed Links
Gene Details
- Gene
- ZFY
- Gene Name
- zinc finger protein, Y-linked
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145275.1 |
2011 |
Missense Mutation |
GAC,GGC |
D527G |
NP_001138747.1 |
NM_001145276.1 |
2011 |
Missense Mutation |
GAC,GGC |
D413G |
NP_001138748.1 |
NM_003411.3 |
2011 |
Missense Mutation |
GAC,GGC |
D604G |
NP_003402.2 |
XM_005262570.2 |
2011 |
Missense Mutation |
GAC,GGC |
D604G |
XP_005262627.1 |
XM_011531473.2 |
2011 |
Missense Mutation |
GAC,GGC |
D578G |
XP_011529775.1 |
XM_011531474.2 |
2011 |
Missense Mutation |
GAC,GGC |
D578G |
XP_011529776.1 |
XM_011531475.2 |
2011 |
Missense Mutation |
GAC,GGC |
D553G |
XP_011529777.1 |
XM_017030075.1 |
2011 |
Missense Mutation |
GAC,GGC |
D604G |
XP_016885564.1 |
XM_017030076.1 |
2011 |
Missense Mutation |
GAC,GGC |
D553G |
XP_016885565.1 |
XM_017030077.1 |
2011 |
Missense Mutation |
GAC,GGC |
D527G |
XP_016885566.1 |
View Full Product Details