Product Details
- SNP ID
-
rs112066282
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:99284581 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTTCCTCCTGCAGGTGAACCTGC[C/T]GGTGCCATCCTGCAGGGTCCCCTGA
- Phenotype
-
MIM: 103700
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADH1A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs28364311] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADH1A
- Gene Name
- alcohol dehydrogenase 1A (class I), alpha polypeptide
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000667.3 |
499 |
Missense Mutation |
AGC,GGC |
S129G |
NP_000658.1 |
- Gene
- LOC100507053
- Gene Name
- uncharacterized LOC100507053
There are no transcripts associated with this gene.
View Full Product Details