Product Details

SNP ID
rs113738337
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:103020269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTGTGAGTTTTTTTTTTTTTTC[C/T]GCATGAAATCTTATGTGGAAAGTTG
Phenotype
MIM: 611527 MIM: 611789
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC9B1 PubMed Links

Gene Details

Gene
SLC9B1
Gene Name
solute carrier family 9 member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100874.2 2368 Intron NP_001094344.1
NM_139173.3 2368 Intron NP_631912.2
XM_006714093.3 2368 Intron XP_006714156.1
XM_011531622.1 2368 Intron XP_011529924.1
XM_011531623.1 2368 Intron XP_011529925.1
Gene
SLC9B2
Gene Name
solute carrier family 9 member B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300754.1 2368 Intron NP_001287683.1
NM_001300756.1 2368 Intron NP_001287685.1
NM_178833.5 2368 Intron NP_849155.2
XM_006714085.2 2368 UTR 3 XP_006714148.1
XM_006714086.3 2368 Intron XP_006714149.1

View Full Product Details