Product Details

SNP ID
rs115231456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:103025715 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAGGTAGACTCACATGGGCCAC[A/T]GAAATAAAGGAACCTGGGTGGTCAG
Phenotype
MIM: 611527 MIM: 611789
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC9B1 PubMed Links

Gene Details

Gene
SLC9B1
Gene Name
solute carrier family 9 member B1
There are no transcripts associated with this gene.

Gene
SLC9B2
Gene Name
solute carrier family 9 member B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300754.1 2364 UTR 3 NP_001287683.1
NM_001300756.1 2364 UTR 3 NP_001287685.1
NM_178833.5 2364 UTR 3 NP_849155.2
XM_006714085.2 2364 Intron XP_006714148.1
XM_006714086.3 2364 UTR 3 XP_006714149.1

View Full Product Details