Product Details

SNP ID
rs114544911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:108624699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAAATCCTTGCTCTTGGTGAGGT[A/G]TCTCTTGCTAATATCCCTTTACCTT
Phenotype
MIM: 616862
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPL34 PubMed Links

Gene Details

Gene
RPL34
Gene Name
ribosomal protein L34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000995.4 Intron NP_000986.2
NM_001319232.1 Intron NP_001306161.1
NM_001319234.1 Intron NP_001306163.1
NM_001319235.1 Intron NP_001306164.1
NM_001319236.1 Intron NP_001306165.1
NM_033625.3 Intron NP_296374.1
Gene
RPL34-AS1
Gene Name
RPL34 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details