Product Details

SNP ID
rs114918376
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:143559680 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGCTTCTAGGAGTAGTGGTATC[A/G]GCGTCTGCTGCGAGAGTCGGGCTCC
Phenotype
MIM: 603375
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GUSBP5 PubMed Links

Gene Details

Gene
GUSBP5
Gene Name
glucuronidase, beta pseudogene 5
There are no transcripts associated with this gene.

Gene
SMARCA5
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 5
There are no transcripts associated with this gene.

View Full Product Details