Product Details

SNP ID
rs116491410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:154536406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGGGGGTTTTCTAATAAGACTG[C/T]CAAAATATTACATCTAGAATAGTTA
Phenotype
MIM: 605961
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLRG1 PubMed Links

Gene Details

Gene
PLRG1
Gene Name
pleiotropic regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201564.1 1922 UTR 3 NP_001188493.1
NM_002669.3 1922 UTR 3 NP_002660.1

View Full Product Details