Product Details

SNP ID
rs113898225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135495273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAATGGCAGGACTTCACGGCCCC[A/G]TTTGCATATTTTCCTACTCCGCAGA
Phenotype
MIM: 614502 MIM: 611971 MIM: 614056
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf116 PubMed Links

Gene Details

Gene
C9orf116
Gene Name
chromosome 9 open reading frame 116
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048265.1 629 UTR 3 NP_001041730.1
NM_144654.2 629 UTR 3 NP_653255.1
Gene
LOC101928525
Gene Name
uncharacterized LOC101928525
There are no transcripts associated with this gene.

Gene
MRPS2
Gene Name
mitochondrial ribosomal protein S2
There are no transcripts associated with this gene.

Gene
PPP1R26
Gene Name
protein phosphatase 1 regulatory subunit 26
There are no transcripts associated with this gene.

View Full Product Details