Product Details

SNP ID
rs116106989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:72130522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTACGGGATCTGGTCTGAGGGA[A/G]CCACTGAGGTGCTCACAGCAGCGAT
Phenotype
MIM: 605207
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP26B1 PubMed Links

Gene Details

Gene
CYP26B1
Gene Name
cytochrome P450 family 26 subfamily B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277742.1 3047 UTR 3 NP_001264671.1
NM_019885.3 3047 UTR 3 NP_063938.1
XM_005264433.4 3047 UTR 3 XP_005264490.1
XM_011532988.1 3047 UTR 3 XP_011531290.1

View Full Product Details