Product Details

SNP ID
rs114797441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:26250377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTTTTCTGTTTTACATGTTACA[A/G]ATATTTTCTCTGAATCTATTTGCCT
Phenotype
MIM: 600890 MIM: 143450
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HADHA PubMed Links

Gene Details

Gene
HADHA
Gene Name
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit
There are no transcripts associated with this gene.

Gene
HADHB
Gene Name
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000183.2 Intron NP_000174.1
NM_001281512.1 Intron NP_001268441.1
NM_001281513.1 Intron NP_001268442.1
XM_011532803.1 Intron XP_011531105.1
XM_017003939.1 Intron XP_016859428.1

View Full Product Details