Product Details

SNP ID
rs115854567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27036226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCTGGAAACATAGGAGCAAGGT[A/C]CTGCTATTATTTCCCATTTTTATAG
Phenotype
MIM: 605788 MIM: 615301
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MAPRE3 PubMed Links

Gene Details

Gene
MAPRE3
Gene Name
microtubule associated protein RP/EB family member 3
There are no transcripts associated with this gene.

Gene
TMEM214
Gene Name
transmembrane protein 214
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083590.1 Intron NP_001077059.1
NM_017727.4 Intron NP_060197.4
XM_005264381.3 Intron XP_005264438.1
XM_005264382.3 Intron XP_005264439.1
XM_005264383.3 Intron XP_005264440.1

View Full Product Details