Product Details

SNP ID
rs114888789
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85839626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAGAAGGGTTGTGACCTTCTCC[A/G]ACCAGCCCAGGCCTCACGCCGCTGC
Phenotype
MIM: 604402
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ST3GAL5 PubMed Links

Gene Details

Gene
ST3GAL5
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042437.1 2018 UTR 3 NP_001035902.1
NM_003896.3 2018 UTR 3 NP_003887.3
XM_005264630.4 2018 UTR 3 XP_005264687.1
XM_011533143.2 2018 UTR 3 XP_011531445.1
XM_017005199.1 2018 UTR 3 XP_016860688.1
XM_017005200.1 2018 UTR 3 XP_016860689.1
XM_017005201.1 2018 UTR 3 XP_016860690.1
XM_017005202.1 2018 UTR 3 XP_016860691.1
XM_017005203.1 2018 UTR 3 XP_016860692.1
XM_017005204.1 2018 UTR 3 XP_016860693.1
XM_017005205.1 2018 UTR 3 XP_016860694.1
XM_017005206.1 2018 UTR 3 XP_016860695.1
XM_017005207.1 2018 UTR 3 XP_016860696.1
XM_017005208.1 2018 UTR 3 XP_016860697.1
XM_017005209.1 2018 UTR 3 XP_016860698.1
XM_017005210.1 2018 UTR 3 XP_016860699.1
XM_017005211.1 2018 UTR 3 XP_016860700.1
XM_017005212.1 2018 UTR 3 XP_016860701.1
XM_017005213.1 2018 UTR 3 XP_016860702.1
XM_017005214.1 2018 UTR 3 XP_016860703.1
XM_017005215.1 2018 UTR 3 XP_016860704.1

View Full Product Details