Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001042437.1 | 1525 | UTR 3 | NP_001035902.1 | ||
NM_003896.3 | 1525 | UTR 3 | NP_003887.3 | ||
XM_005264630.4 | 1525 | UTR 3 | XP_005264687.1 | ||
XM_011533143.2 | 1525 | UTR 3 | XP_011531445.1 | ||
XM_017005199.1 | 1525 | UTR 3 | XP_016860688.1 | ||
XM_017005200.1 | 1525 | UTR 3 | XP_016860689.1 | ||
XM_017005201.1 | 1525 | UTR 3 | XP_016860690.1 | ||
XM_017005202.1 | 1525 | UTR 3 | XP_016860691.1 | ||
XM_017005203.1 | 1525 | UTR 3 | XP_016860692.1 | ||
XM_017005204.1 | 1525 | UTR 3 | XP_016860693.1 | ||
XM_017005205.1 | 1525 | UTR 3 | XP_016860694.1 | ||
XM_017005206.1 | 1525 | UTR 3 | XP_016860695.1 | ||
XM_017005207.1 | 1525 | UTR 3 | XP_016860696.1 | ||
XM_017005208.1 | 1525 | UTR 3 | XP_016860697.1 | ||
XM_017005209.1 | 1525 | UTR 3 | XP_016860698.1 | ||
XM_017005210.1 | 1525 | UTR 3 | XP_016860699.1 | ||
XM_017005211.1 | 1525 | UTR 3 | XP_016860700.1 | ||
XM_017005212.1 | 1525 | UTR 3 | XP_016860701.1 | ||
XM_017005213.1 | 1525 | UTR 3 | XP_016860702.1 | ||
XM_017005214.1 | 1525 | UTR 3 | XP_016860703.1 | ||
XM_017005215.1 | 1525 | UTR 3 | XP_016860704.1 |