Product Details

SNP ID
rs111609268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27664393 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCTCGGTACCACGCAGTGCTGC[A/G]GCACAGGGCGTCCGGCCCTGACGGA
Phenotype
MIM: 602655 MIM: 612762
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC4A1AP PubMed Links

Gene Details

Gene
SLC4A1AP
Gene Name
solute carrier family 4 member 1 adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018158.2 923 Missense Mutation CAG,CGG Q214R NP_060628.2
Gene
SUPT7L
Gene Name
SPT7-like STAGA complex gamma subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282729.1 923 Intron NP_001269658.1
NM_001282730.1 923 Intron NP_001269659.1
NM_001282731.1 923 Intron NP_001269660.1
NM_001282732.1 923 Intron NP_001269661.1
NM_014860.2 923 Intron NP_055675.1
XM_005264672.4 923 Intron XP_005264729.1

View Full Product Details