Product Details
- SNP ID
-
rs116815829
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:28894916 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGCCTGGGCGCCAGCGCGGCTGC[A/G]GGCCAAGGTAAAGCGAGCGGGACTG
- Phenotype
-
MIM: 616195
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WDR43
PubMed Links
Gene Details
- Gene
- WDR43
- Gene Name
- WD repeat domain 43
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015131.2 |
274 |
Missense Mutation |
CAG,CGG |
Q73R |
NP_055946.1 |
View Full Product Details