Product Details

SNP ID
rs113852987
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:69021614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTGAAAAACTCCAAATTCTTCA[A/G]GATGGTGTACTTGTATCTATATTCT
Phenotype
MIM: 614789 MIM: 601126
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EOGT PubMed Links

Gene Details

Gene
EOGT
Gene Name
EGF domain specific O-linked N-acetylglucosamine transferase
There are no transcripts associated with this gene.

Gene
TMF1
Gene Name
TATA element modulatory factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007114.2 5092 UTR 3 NP_009045.2
XM_011534056.2 5092 UTR 3 XP_011532358.1

View Full Product Details