Product Details

SNP ID
rs114401001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:43079503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTTCAGGATGGGAGAAGGGGAA[A/G]TCAGGTCCCTTATCTCTAGGGCAAA
Phenotype
MIM: 614828
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POMGNT2 PubMed Links

Gene Details

Gene
POMGNT2
Gene Name
protein O-linked mannose N-acetylglucosaminyltransferase 2 (beta 1,4-)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032806.5 2282 UTR 3 NP_116195.2
XM_005265515.2 2282 UTR 3 XP_005265572.1
XM_011534163.2 2282 UTR 3 XP_011532465.1
XM_011534164.1 2282 UTR 3 XP_011532466.1
XM_017007353.1 2282 UTR 3 XP_016862842.1

View Full Product Details