Product Details

SNP ID
rs115643713
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:64094458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGCAGATTTTTCTCTATCCTAT[G/T]GATTCATCGTATTCAAAACACAGGA
Phenotype
MIM: 608501
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
LINC00994 PubMed Links

Gene Details

Gene
LINC00994
Gene Name
long intergenic non-protein coding RNA 994
There are no transcripts associated with this gene.

Gene
PRICKLE2
Gene Name
prickle planar cell polarity protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198859.3 7438 UTR 3 NP_942559.1
XM_011533432.2 7438 UTR 3 XP_011531734.1
XM_011533433.2 7438 UTR 3 XP_011531735.1
XM_011533434.2 7438 UTR 3 XP_011531736.1
XM_011533435.2 7438 UTR 3 XP_011531737.1
XM_011533436.2 7438 UTR 3 XP_011531738.1
XM_011533437.2 7438 UTR 3 XP_011531739.1
XM_011533438.2 7438 UTR 3 XP_011531740.1
XM_011533440.2 7438 UTR 3 XP_011531742.1
XM_017005798.1 7438 UTR 3 XP_016861287.1
XM_017005799.1 7438 UTR 3 XP_016861288.1
Gene
PRICKLE2-AS1
Gene Name
PRICKLE2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
PRICKLE2-AS2
Gene Name
PRICKLE2 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details