Product Details

SNP ID
rs116740186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56648169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCCAGTTGGAGTTTCTGTCTTC[C/T]GGCCCAACAGGCTTCTTTCCAAGTT
Phenotype
MIM: 607275
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOPX PubMed Links

Gene Details

Gene
HOPX
Gene Name
HOP homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145459.1 1240 UTR 3 NP_001138931.1
NM_001145460.1 1240 UTR 3 NP_001138932.1
NM_032495.5 1240 UTR 3 NP_115884.4
NM_139211.4 1240 UTR 3 NP_631957.1
NM_139212.3 1240 UTR 3 NP_631958.1
XM_006714052.2 1240 Intron XP_006714115.1
XM_017008728.1 1240 UTR 3 XP_016864217.1
XM_017008729.1 1240 UTR 3 XP_016864218.1
XM_017008730.1 1240 UTR 3 XP_016864219.1
XM_017008731.1 1240 UTR 3 XP_016864220.1
XM_017008732.1 1240 UTR 3 XP_016864221.1
XM_017008733.1 1240 UTR 3 XP_016864222.1
XM_017008734.1 1240 UTR 3 XP_016864223.1

View Full Product Details