Product Details

SNP ID
rs112571867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56649650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACAGAAGTGACCATGGATGTAC[A/G]GAAGGCCCCAGATGGGATGTTTCTA
Phenotype
MIM: 607275
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HOPX PubMed Links

Gene Details

Gene
HOPX
Gene Name
HOP homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145459.1 1891 Intron NP_001138931.1
NM_001145460.1 1891 Intron NP_001138932.1
NM_032495.5 1891 Intron NP_115884.4
NM_139211.4 1891 Intron NP_631957.1
NM_139212.3 1891 Intron NP_631958.1
XM_006714052.2 1891 UTR 3 XP_006714115.1
XM_017008728.1 1891 Intron XP_016864217.1
XM_017008729.1 1891 Intron XP_016864218.1
XM_017008730.1 1891 Intron XP_016864219.1
XM_017008731.1 1891 Intron XP_016864220.1
XM_017008732.1 1891 Intron XP_016864221.1
XM_017008733.1 1891 Intron XP_016864222.1
XM_017008734.1 1891 Intron XP_016864223.1

View Full Product Details