Product Details

SNP ID
rs114522539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:172957715 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTATTCTAAGAAGAACCTCTGAC[C/T]TTTACTGTGTGTGCGCCGTTATGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERGIC1 PubMed Links

Gene Details

Gene
ERGIC1
Gene Name
endoplasmic reticulum-golgi intermediate compartment 1
There are no transcripts associated with this gene.

Gene
LOC100268168
Gene Name
uncharacterized LOC100268168
There are no transcripts associated with this gene.

Gene
RPL26L1
Gene Name
ribosomal protein L26 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317980.1 Intron NP_001304909.1
NM_001317981.1 Intron NP_001304910.1
NM_001317982.1 Intron NP_001304911.1
NM_016093.3 Intron NP_057177.1
XM_005265921.3 Intron XP_005265978.1
XM_011534565.2 Intron XP_011532867.1
XM_017009519.1 Intron XP_016865008.1
XM_017009520.1 Intron XP_016865009.1

View Full Product Details