Product Details

SNP ID
rs116113799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137299960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACACAAAGTTTTATAAAAAGCGA[C/G]GGAGGGCTGCCCGAATACGACCAGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EXD3 PubMed Links

Gene Details

Gene
EXD3
Gene Name
exonuclease 3'-5' domain containing 3
There are no transcripts associated with this gene.

Gene
NRARP
Gene Name
NOTCH-regulated ankyrin repeat protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004354.2 2292 UTR 3 NP_001004354.1

View Full Product Details