Product Details

SNP ID
rs115566777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:132844112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGCCACAGCAGTAACACAGAAC[A/G]GTGTCTCCCGTGACTCTGTAATCTG
Phenotype
MIM: 605209
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHFR PubMed Links

Gene Details

Gene
CHFR
Gene Name
checkpoint with forkhead and ring finger domains, E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161344.1 1755 Silent Mutation ACC,ACT T598T NP_001154816.1
NM_001161345.1 1755 Silent Mutation ACC,ACT T597T NP_001154817.1
NM_001161346.1 1755 Silent Mutation ACC,ACT T586T NP_001154818.1
NM_001161347.1 1755 Silent Mutation ACC,ACT T506T NP_001154819.1
NM_018223.2 1755 Silent Mutation ACC,ACT T557T NP_060693.2

View Full Product Details