Product Details
- SNP ID
-
rs116011732
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:132922655 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACAATTAATAAAGGCACTATTTA[C/T]AAATATGTGTTCACAATTAAAGACA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF605
PubMed Links
Gene Details
- Gene
- ZNF605
- Gene Name
- zinc finger protein 605
View Full Product Details