Product Details

SNP ID
rs114214898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:39347347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCCTCTGGTAAACACCTAATA[A/C]ATAATTGTTGAATGAAAGGCAAGGT
Phenotype
MIM: 606710
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LHFP PubMed Links

Gene Details

Gene
LHFP
Gene Name
lipoma HMGIC fusion partner
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005780.2 Intron NP_005771.1
XM_011534861.1 Intron XP_011533163.1

View Full Product Details