Product Details

SNP ID
rs117029125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:62525820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTGCGTGTGCTGATGGTAAAAG[C/T]CTCCTCCACTCTTCAGCCTACTCGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZCCHC2 PubMed Links

Gene Details

Gene
ZCCHC2
Gene Name
zinc finger CCHC-type containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017742.5 1001 Intron NP_060212.4
XM_006722493.3 1001 Intron XP_006722556.1
XM_011526044.1 1001 UTR 5 XP_011524346.1
XM_017025802.1 1001 Intron XP_016881291.1

View Full Product Details