Product Details

SNP ID
rs116398727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:143884307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCTCTTGAATTTTACACAGATG[A/G]CAATCAGGAATTGTTTCCCTGTGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZC2HC1B PubMed Links

Gene Details

Gene
ZC2HC1B
Gene Name
zinc finger C2HC-type containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013623.2 136 Missense Mutation GAC,GGC D11G NP_001013645.1

View Full Product Details