Product Details

SNP ID
rs117863331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:147218873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTTCATTCTTTCTGATAGCTGC[A/G]TAGTTGATCATGTTATCAATGAACA
Phenotype
MIM: 604586
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STXBP5 PubMed Links

Gene Details

Gene
STXBP5
Gene Name
syntaxin binding protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127715.2 Intron NP_001121187.1
NM_139244.4 Intron NP_640337.3
XM_006715339.2 Intron XP_006715402.1
XM_011535453.1 Intron XP_011533755.1
XM_017010267.1 Intron XP_016865756.1
XM_017010268.1 Intron XP_016865757.1

View Full Product Details