Product Details

SNP ID
rs118170485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:148197142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAGGCAGCATTTAGGGAAAATGA[A/G]TCTGGCAAGGTAGATTGGATTCTGA
Phenotype
MIM: 607955
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SASH1 PubMed Links

Gene Details

Gene
SASH1
Gene Name
SAM and SH3 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015278.3 Intron NP_056093.3
XM_005266875.1 Intron XP_005266932.1
XM_011535640.1 Intron XP_011533942.1
XM_017010598.1 Intron XP_016866087.1
XM_017010599.1 Intron XP_016866088.1
XM_017010600.1 Intron XP_016866089.1
XM_017010601.1 Intron XP_016866090.1
XM_017010602.1 Intron XP_016866091.1
XM_017010603.1 Intron XP_016866092.1
XM_017010604.1 Intron XP_016866093.1
XM_017010605.1 Intron XP_016866094.1
XM_017010606.1 Intron XP_016866095.1
XM_017010607.1 Intron XP_016866096.1

View Full Product Details