Product Details

SNP ID
rs116902858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149869273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAGGCCGTTCTTAGAGGAAGGA[C/T]GTGGTAGTAACTGGGGAAAATCCTG
Phenotype
MIM: 609243
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRP11 PubMed Links

Gene Details

Gene
LRP11
Gene Name
LDL receptor related protein 11
There are no transcripts associated with this gene.

Gene
RAET1E
Gene Name
retinoic acid early transcript 1E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243325.1 2016 Intron NP_001230254.1
NM_001243327.1 2016 Intron NP_001230256.1
NM_001243328.1 2016 Intron NP_001230257.1
NM_139165.2 2016 Intron NP_631904.1
XM_011535475.2 2016 Intron XP_011533777.1
XM_011535476.2 2016 Intron XP_011533778.1
XM_011535478.2 2016 Intron XP_011533780.1
XM_011535479.2 2016 Intron XP_011533781.1
XM_017010285.1 2016 UTR 3 XP_016865774.1
XM_017010286.1 2016 UTR 3 XP_016865775.1
XM_017010287.1 2016 UTR 3 XP_016865776.1
XM_017010288.1 2016 Intron XP_016865777.1
Gene
RAET1E-AS1
Gene Name
RAET1E antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details