Product Details
- SNP ID
-
rs114441771
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:160532622 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTCTCAATAACAAGGAGCTGGGCT[G/T]CCTTGAGAAGGCCAGTCCCAAAGGT
- Phenotype
-
MIM: 152200
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LPA
PubMed Links
Gene Details
- Gene
- LPA
- Gene Name
- lipoprotein(a)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005577.2 |
5990 |
Missense Mutation |
GAA,GCA |
E1957A |
NP_005568.2 |
View Full Product Details