Product Details
- SNP ID
-
rs115890909
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:162728307 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATGCCGAAGAGGACCAAGCTGCT[A/G]GCACAACAGCCGCTCCCGGTGCACC
- Phenotype
-
MIM: 608427
MIM: 602544
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PACRG
PubMed Links
Gene Details
- Gene
- PACRG
- Gene Name
- PARK2 coregulated
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080378.1 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
NP_001073847.1 |
NM_001080379.1 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
NP_001073848.1 |
NM_152410.2 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
NP_689623.2 |
XM_005266825.4 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
XP_005266882.1 |
XM_005266826.4 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
XP_005266883.1 |
XM_006715344.3 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
XP_006715407.1 |
XM_006715345.3 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
XP_006715408.1 |
XM_011535461.2 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
XP_011533763.1 |
XM_011535462.2 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
XP_011533764.1 |
XM_011535466.1 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
XP_011533768.1 |
XM_011535468.2 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
XP_011533770.1 |
XM_011535469.2 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
XP_011533771.1 |
XM_011535471.1 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
XP_011533773.1 |
XM_017010275.1 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
XP_016865764.1 |
XM_017010276.1 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
XP_016865765.1 |
XM_017010277.1 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
XP_016865766.1 |
XM_017010278.1 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
XP_016865767.1 |
XM_017010279.1 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
XP_016865768.1 |
XM_017010280.1 |
221 |
Silent Mutation |
CTA,CTG |
L24L |
XP_016865769.1 |
XM_017010281.1 |
221 |
Intron |
|
|
XP_016865770.1 |
XM_017010282.1 |
221 |
Intron |
|
|
XP_016865771.1 |
- Gene
- PARK2
- Gene Name
- parkin RBR E3 ubiquitin protein ligase
View Full Product Details