Product Details

SNP ID
rs115435591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:165329764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTTGCTTTTGTTGTGGTGAGAAC[A/G]AGTCACCTGGGGCACCACTGTAAAC
Phenotype
MIM: 610652
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDE10A PubMed Links

Gene Details

Gene
PDE10A
Gene Name
phosphodiesterase 10A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130690.2 5591 UTR 3 NP_001124162.1
NM_006661.3 5591 UTR 3 NP_006652.1
XM_006715321.3 5591 UTR 3 XP_006715384.1
XM_011535387.2 5591 UTR 3 XP_011533689.2
XM_011535388.2 5591 UTR 3 XP_011533690.1
XM_011535393.2 5591 UTR 3 XP_011533695.1
XM_017010194.1 5591 UTR 3 XP_016865683.1
XM_017010195.1 5591 UTR 3 XP_016865684.1
XM_017010196.1 5591 UTR 3 XP_016865685.1
XM_017010197.1 5591 Intron XP_016865686.1

View Full Product Details